Learn More
Medchemexpress LLC Phospho-AMPK alpha 2(Ser345) Antibody (YA225) | Protein A affinity purified | 62 kDa | 50 UL

Supplier: Medchemexpress LLC HYP8045250UL
Phospho-AMPK alpha 2(Ser345) Antibody (YA225) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to Phospho-AMPK alpha 2(Ser345). It is for research use only.
- Host: Rabbit
- Clonality: Recombinant, monoclonal
- Species reactivity: Human, mouse
- Purity: Protein A affinity purified
- Conjugation: Non-conjugated
- Modification: Phosphorylated
- Isotype: IgG
- Appearance: Liquid
- Formulation: Supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol. Preservative: 0.05% Sodium Azide.
- Storage & stability: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
- Shipping: Shipped with blue ice.
- Subcellular localization: Cytoplasm; nucleus
- Immunogen: Synthetic phosphopeptide corresponding to residues surrounding Ser345 of Human AMPK alpha 2. AA range: 338-352.
- Gene ID: 5563 [NCBI]
- SwissProt ID: P54646
- RRID: AB_3102528
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.