Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-Beta Catenin (Ser33/Ser37) Antibody (YA3096) | 50 UL
SDP

Supplier:  Medchemexpress LLC HYP8335150UL

Encompass_Preferred

This product is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting Phospho-beta Catenin (Ser33/Ser37). It is designed for research use, providing a specific tool for the detection of phosphorylated beta Catenin in various applications.

  • Rabbit-derived host
  • Recombinant, monoclonal antibody
  • Targets phospho-beta catenin (Ser33/Ser37)
  • Reacts with human and rat species
  • Observed molecular weight of 92 kDa
  • Purified by affinity chromatography
  • Non-conjugated IgG isotype
  • Formulated for stability and performance
  • Store at -20°C for up to 1 year
  • Avoid repeated freeze/thaw cycles
  • Suitable for Western blot applications

Catalog No. 50-004-44965


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.