Learn More
Medchemexpress LLC Phospho-CDC6 (Ser54) Antibody (YA1017)(PBS only) | 10 UL

Supplier: Medchemexpress LLC HYP81295B10UL
Phospho-CDC6 (Ser54) Antibody (YA1017)(PBS only) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Phospho-CDC6 (Ser54). This antibody is supplied in PBS, pH 7.4, and is suitable for applications such as Western Blot (WB) and Immunocytochemistry/Immunofluorescence (ICC/IF).
- Host: Rabbit
- Reactivity: Human
- Clonality No.: YA1017
- SwissProt ID: Q99741
- Gene ID: 990
- Molecular weight: 63 kDa
- Appearance: Liquid
- Conjugation: Non-conjugated
- Applications: WB, ICC/IF
- Formulation: Supplied in PBS, pH 7.4
- Storage at -20°C for 1 year; avoid repeated freeze / thaw cycles
- Shipping with blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.