Learn More
Medchemexpress LLC Phospho-CDK1 (Tyr15) Antibody | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8079610UL
Encompass_Preferred
Phospho-CDK1 (Tyr15) Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed for research applications. It specifically targets Phospho-CDK1 (Tyr15) and is suitable for use in Western Blotting experiments. This antibody is affinity purified and reactive with human, mouse, and rat samples.
  • Rabbit-derived polyclonal antibody
  • Targets Phospho-CDK1 (Tyr15)
  • Reacts with human, mouse, and rat
  • Affinity purified
  • Suitable for Western Blot (WB) applications
  • Supplied in PBS (pH 7.3), 50% glycerol, and 0.5% BSA with 0.02% sodium azide as a preservative
  • Predicted and observed band size of 34 kDa
  • Stored at -20°C for 1 year
  • Ships with blue ice
Catalog No. 50-004-42665