Learn More
Sino Biological Phospho-CREB (Ser133) Antibody, Rabbit MAb 100 µL
Supplier:  Sino Biological 110398R0256100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to residues around Ser133 of Human Phospho-CREB
Catalog No. 50-007-51187