Learn More
Medchemexpress LLC Phospho-FOXO3A (Ser253) antibody (YA197)(PBS only) | 50 UL

Supplier: Medchemexpress LLC HYP8081750UL
Phospho-FOXO3A (Ser253) antibody (YA197) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Phospho-FOXO3A (Ser253). It is supplied in PBS, pH 7.4, and is intended for research use only. This antibody is affinity purified.
- Rabbit-derived, non-conjugated IgG monoclonal antibody
- Targets Phospho-FOXO3A (Ser253)
- Supplied in PBS, pH 7.4
- For research use only
- Predicted molecular weight: 71 kDa; Observed molecular weight: 82 kDa
- Store at -20°C for 1 year
- Avoid repeated freeze/thaw cycles
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.