Learn More
Medchemexpress LLC Phospho-GSK3 beta (Ser 9) Antibody (YA192) | Protein A affinity purified | 47 kDa | 10 UL

Supplier: Medchemexpress LLC HYP8046210UL
Phospho-GSK3 beta (Ser 9) Antibody (YA192) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Phospho-GSK3 beta (Ser 9). GSK-3 beta is a constitutively active protein kinase involved in regulating diverse cellular processes, including glucose homeostasis, Wnt signaling, and transcription factors. It also plays crucial roles in microtubule dynamics, NF-kappa-B regulation, pancreatic beta-cell replication, apoptosis control, circadian rhythms, autophagy, and extrinsic apoptotic signaling.
- Targets Phospho-GSK3 beta (Ser 9)
- Rabbit-derived monoclonal antibody
- Reactivity with human samples
- Protein A affinity purified
- Applications include Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Immunohistochemistry-Paraffin (IHC-P)
- Supplied in TBS, BSA, and Glycerol with Sodium Azide as a preservative
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.