Learn More
ABclonal Technology Phospho-GSK3B-S9 Mouse mAb
SDP
Supplier:  ABclonal Technology AP125820UL
Encompass_Preferred
The protein encoded by this gene is a serine-threonine kinase belonging to the glycogen synthase kinase subfamily It is a negative regulator of glucose homeostasis and is involved in energy metabolism inflammation ER-stress mitochondrial dysfunction and apoptotic pathways Defects in this gene have been associated with Parkinson disease and Alzheimer disease
Catalog No. 50-000-14148