Learn More
Medchemexpress LLC Phospho-IGF1 Receptor (Tyr1166) Antibody (YA186) | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP8082550UL
Encompass_Preferred
This Phospho-IGF1 Receptor (Tyr1166) Antibody (YA186) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target Phospho-IGF1 Receptor (Tyr1166). It exhibits reactivity with human, mouse, and rat samples and is supplied as a liquid. The antibody is affinity purified and suitable for applications such as Western Blot (WB) and Immunoprecipitation (IP). The IGF1 Receptor plays a critical role in cellular transformation, possesses tyrosine kinase activity, and acts as an anti-apoptotic agent by enhancing cell survival, often overexpressed in malignant tissues.
  • Designed for targeting Phospho-IGF1 Receptor (Tyr1166)
  • Reactive with human, mouse, and rat samples
  • Suitable for Western Blot (WB) at 1:500-1:1,000 dilution
  • Suitable for Immunoprecipitation (IP) at 1:20 dilution
  • Affinity purified
  • Store at -20°C for 1 year; avoid repeated freeze/thaw cycles
Catalog No. 50-004-42023