Learn More
ABclonal Technology Phospho-MDM2-S166 Rabbit mAb
SDP
Supplier:  ABclonal Technology AP1308100UL
Encompass_Preferred
This gene encodes a nuclear-localized E3 ubiquitin ligase The encoded protein can promote tumor formation by targeting tumor suppressor proteins such as p53 for proteasomal degradation This gene is itself transcriptionally-regulated by p53 Overexpression or amplification of this locus is detected in a variety of different cancers There is a pseudogene for this gene on chromosome 2 Alternative splicing results in a multitude of transcript variants many of which may be expressed only in tumor cells
Catalog No. 50-000-14162