Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-NF-KB p65 (Ser536) Antibody | 100 UL
SDP

Supplier:  Medchemexpress LLC HYP80839100UL

Encompass_Preferred

Phospho-NF-KB p65 (Ser536) Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed to target Phospho-NF-KB p65 (Ser536). NF-kappa-B is a crucial transcription factor involved in various biological processes including inflammation, immunity, differentiation, cell growth, and apoptosis.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Reactivity: Human, Mouse, Rat
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Modification: Phosphorylated
  • Formulation: Supplied in 1*PBS (pH 7.3), 50% glycerol, 0.5% BSA, and 0.02% sodium azide
  • Appearance: Liquid
  • Storage & stability: Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Sensitivity: Endogenous
  • Applications: WB, IHC-P, ICC/IF

Catalog No. 50-004-42892


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.