Learn More
Medchemexpress LLC Phospho-NF-KB p65 (Thr254) Antibody | 10 UL

Supplier: Medchemexpress LLC HYP8084010UL
NF-kappa-B is a ubiquitous transcription factor involved in several biological processes. It is held in the cytoplasm in an inactive state by specific inhibitors. Upon degradation of the inhibitor, NF-kappa-B moves to the nucleus and activates transcription of specific genes. The most abundant form of NF-kappa-B is NFKB1 complexed with RELA.
- Rabbit-derived, non-conjugated IgG polyclonal antibody
- Targets Phospho-NF-KB p65 (Thr254)
- Reactivity: Human, mouse, rat
- SwissProt ID: Q04206
- Research field: Cell biology
- Formulation: Supplied in 1*PBS (pH 7.3), 50% glycerol and 0.5% BSA with 0.02% sodium azide
- Appearance: Liquid
- Applications: WB (1:500-1:1000), IHC-P (1:50-1:100), ICC/IF (1:50-1:200)
- Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
- Shipping: Shipped with blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.