Learn More
Medchemexpress LLC Phospho-p27 KIP 1 (Ser10) Antibody (YA1494) (PBS only) | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP81749A50UL
Encompass_Preferred
Phospho-p27 KIP 1 (Ser10) Antibody (YA1494) is a Rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Phospho-p27 KIP 1 (Ser10). Supplied in PBS, pH 7.4, this antibody is crucial for regulating cell cycle progression by inhibiting CDK2 activity and is involved in G1 arrest. It can act as an inhibitor or activator of cyclin type D-CDK4 complexes depending on its phosphorylation state. Expressed across various tissues, its levels are maximal in quiescent cells and early G1, decreasing as cells approach S-phase.
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated, targets Phospho-p27 KIP 1 (Ser10)
  • Regulates cell cycle progression and G1 arrest
  • Inhibits CDK2 activity
  • Expressed in all tissues tested
Catalog No. 50-004-41416