Learn More
Medchemexpress LLC Phospho-p38 alpha/MAPK14 (Thr180+Tyr182) Antibody | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP80776A10UL
Encompass_Preferred
Phospho-p38 alpha/MAPK14 (Thr180+Tyr182) Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody specifically targeting phosphorylated p38 alpha/MAPK14. This antibody is developed for research use only and is suitable for various immunological assays. It comes as an affinity-purified liquid formulation with a concentration of 1 mg/mL.
  • Rabbit-derived IgG polyclonal antibody
  • Targets specifically phosphorylated p38 alpha/MAPK14 (Thr180+Tyr182)
  • Affinity purified for high specificity
  • Suitable for multiple applications including Western Blot, IHC-P, FC, ICC/IF, and ELISA
  • Reacts with human and mouse samples
  • Stable for 1 year when stored at -20°C
Catalog No. 50-004-42849