Learn More
Medchemexpress LLC Phospho-p53 (Ser6) Antibody (YA166) | 50 UL

Supplier: Medchemexpress LLC HYP8084450UL
The Phospho-p53 (Ser6) Antibody (YA166) is a rabbit-derived, non-conjugated IgG monoclonal antibody developed for research use. It specifically targets phosphorylated Ser6 of human p53, a critical tumor suppressor protein involved in cell cycle arrest, DNA repair, and apoptosis. The antibody is supplied as a liquid formulation in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.05% BSA, and 0.01% Sodium azide.
- Host: Rabbit
- Clonality: Recombinant, monoclonal
- Species reactivity: Human
- Purity: Affinity purified
- Conjugation: Non-conjugated
- Isotype: IgG
- Appearance: Liquid
- Applications: Western blot, immunohistochemistry-paraffin, immunoprecipitation
- Storage: Store at -20°C for 1 year, avoid repeated freeze/thaw cycles
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.