Learn More
Medchemexpress LLC Phospho-p95/NBS1 (Ser343) Antibody (YA165) | 50 UL

Supplier: Medchemexpress LLC HYP8047350UL
Phospho-p95/NBS1 (Ser343) Antibody (YA165) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to Phospho-p95/NBS1 (Ser343).
- Application: WB, ICC/IF, IHC-P, IP
- Reactivity: Human, Mouse
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.