Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-PDGFR beta (Tyr740) Antibody (YA3062) | 50 UL
SDP

Supplier:  Medchemexpress LLC HYP8331750UL

Encompass_Preferred

Phospho-PDGFR beta (Tyr740) Antibody (YA3062) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Phospho-PDGFR beta (Tyr740). This antibody is essential for understanding its roles in embryonic development, cell proliferation, survival, differentiation, chemotaxis, and migration, and is widely used in cardiovascular research.

  • Host: Rabbit
  • Reactivity: Mouse, rat
  • Conjugation: Non-conjugated
  • Research field: Cardiovascular
  • Applications: WB (dilution ratio: 1:500-1:1000), ICC/IF (dilution ratio: 1:50-1:200)
  • Predicted band size: 124 kDa
  • Observed band size: 190 kDa
  • Supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Store at -20°C for 1 year; avoid repeated freeze / thaw cycles

Catalog No. 50-004-44984


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.