Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-PI3 Kinase p110 beta (Ser1070) Antibody | 50 UL
SDP

Supplier:  Medchemexpress LLC HYP8121150UL

Encompass_Preferred

Phospho-PI3 Kinase p110 beta (Ser1070) antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody targeting phospho-PI3 Kinase p110 beta (Ser1070). This antibody is designed for research use only and is involved in essential cellular processes including cell growth, survival, proliferation, motility, and morphology by activating signaling cascades. It is formulated in 0.01M TBS (pH7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.

  • Host: Rabbit
  • Clonality: Polyclonal
  • Predicted band size: 110-123 kDa
  • Observed band size: 110 kDa
  • Species reactivity: Human, mouse, rat
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Storage: Stored at -20°C for 1 year
  • Applications: WB, IHC-P, IHC-F, ICC/IF

Catalog No. 50-004-42921


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.