Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-PI3 Kinase p85/p55 (Tyr467/Tyr199) Antibody | 10 UL
SDP

Supplier:  Medchemexpress LLC HYP8084610UL

Encompass_Preferred

This Phospho-PI3 Kinase p85/p55 (Tyr467/Tyr199) Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody for research use. It targets Phospho-PI3 Kinase p85/p55 (Tyr467/Tyr199) and reacts with human, mouse, and rat species. It is affinity purified and modified by phosphorylation.

  • Supplied in 1*PBS (pH 7.3), 50% glycerol, 0.5% BSA, and 0.02% sodium azide.
  • Immunogen: synthetic phosphopeptide for Tyr467/199 of human PI3-kinase p85-alpha/gamma.
  • Store at -20°C for 1 year; avoid freeze/thaw cycles.
  • Shipped with blue ice.
  • Involved in signaling responses to growth factors and receptors.
  • Modulates ER stress response via XBP1 isoform 2 nuclear translocation.
  • Undergoes polyubiquitination and phosphorylation due to signaling.
  • Forms a heterodimer with a p110 catalytic subunit, interacting with various proteins.

Catalog No. 50-004-42207


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.