Learn More
Medchemexpress LLC Phospho-PI3 kinase p85/p55 (Tyr467/Tyr199) antibody | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP8084650UL
Encompass_Preferred
Phospho-PI3 Kinase p85/p55 (Tyr467/Tyr199) Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed for research use. It targets Phospho-PI3 Kinase p85/p55 (Tyr467/Tyr199) and is suitable for various applications including Western Blot, immunohistochemistry, and immunocytochemistry.
  • Rabbit-derived polyclonal antibody
  • Targets phospho-PI3 kinase p85/p55 (Tyr467/Tyr199)
  • Suitable for multiple applications (WB, IHC-P, IHC-F, ICC/IF, ELISA)
  • Affinity purified
  • Formulated in 1xPBS (pH 7.3), 50% glycerol, and 0.5% BSA with 0.02% sodium azide
  • Stable for 1 year when stored at -20°C
  • Species reactivity: Human, mouse, rat
Catalog No. 50-004-42096