Learn More
Medchemexpress LLC Phospho-PLK1 (Thr210) Antibody (YA161) | 68 kDa | 100UL

Supplier: Medchemexpress LLC HYP80474100UL
Phospho-PLK1 (Thr210) Antibody (YA161) is a rabbit-derived, non-conjugated IgG monoclonal antibody. It specifically targets Phospho-PLK1 at the Thr210 phosphorylation site.
- Suitable for Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC) applications.
- Reacts with human samples.
- Predicted and observed molecular weight of 68 kDa.
- Formulated in 1xTBS (pH 7.4) with 0.05% BSA, 40% glycerol, and 0.05% sodium azide.
- Stable for 1 year when stored at -20°C.
- Appearance: liquid.
- Concentration: 1 mg/mL.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.