Learn More
Cell Signaling Technology Phospho-S6 Ribosomal Protein
SDP
Supplier:  Cell Signaling Technology 15448S
Encompass
Phospho-S6 Ribosomal Protein (Ser235/236) (D57.2.2E) XP(R) Rabbit mAb (mFluor(tm) Red 780 Conjugate) 100 ul
Catalog No. 50-272-8452