Learn More
Medchemexpress LLC Phospho-TAK1 (Ser439) Antibody (YA144) | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8085810UL
Encompass_Preferred
This Phospho-TAK1 (Ser439) Antibody (YA144) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Phospho-TAK1 (Ser439). It is affinity purified with a molecular weight of 67-78 kDa. Intended for research use only, it is stored at -20°C for up to one year, with precautions against repeated freeze/thaw cycles.
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Specifically targets Phospho-TAK1 (Ser439)
  • Affinity purified
  • Molecular weight of 67-78 kDa
  • For research use only
  • Stable for 1 year when stored at -20°C
  • Avoid repeated freeze/thaw cycles
Catalog No. 50-004-42644