Learn More
BIOHIPPO Phospho-Tyrosine Mouse mAb
SDP
Supplier:  BIOHIPPO BHA21821306
Encompass
U-Blot Pan-Phospho-Tyrosine Mouse mAb is a Mouse-derived monoclonal antibody that specifically recognizes Phospho-Tyrosine a tyrosine phosphorylation modification at Tyr(Y) It shows reactivity with Species Independent Validated applications include WB IHC-P ICC/IF FC Recommended dilutions WB 11000 IHC-P 1100-1200 ICC/IF 1100-1200 FC 1100-1200 Purified by protein a Phospho-Tyrosine (pY) is a widely occurring post-translational modification on tyrosine residues in eukaryotic cells This modification is catalyzed by protein tyrosine kinases (PTKs) which covalently attach a phosphate group to the phenolic hydroxyl group of tyrosine and can be reversibly removed by protein tyrosine phosphatases (PTPs) Tyrosine phosphorylation serves as a core mechanism in cellular signal transduction regulating cell proliferation differentiation apoptosis migration and metabolism Supplied as 100 uL
Catalog No. 50-009-75557