Learn More
Cayman Chemical PhosphoenolpyruvIc AcIdpota 1g
SDP
Supplier:  Cayman Chemical 191921 G
Encompass_Preferred
An enzyme substrate for glycolysis and gluconeogenesis pathways; metabolized to pyruvate by pyruvate kinase during glycolysis; formed from phosphoenolpyruvate carboxykinase-catalyzed oxaloacetate decarboxylation during gluconeogenesis
Catalog No. 50-298-3712