Learn More
Medchemexpress LLC Phosphoglucomutase, Rabbit muscle | 9001-81-4 | approximately 62,000 Daltons | 500 U
SDP
Supplier:  Medchemexpress LLC HYP2820500U
Encompass_Preferred
Phosphoglucomutase, Rabbit muscle, is an enzyme frequently utilized in biochemical studies. This enzyme facilitates the transfer of a phosphate group on the α-D-glucose monomer, promoting the interconversion of glucose-1-phosphate and glucose-6-phosphate. It plays a significant role in glycolysis and gluconeogenesis, important in the metabolism of proteins, lipids, and nucleic acids.
  • Optimum pH: 7.5-8.0, with activity maintained within the range of 4.5-8.5
  • Activators include: Divalent cations (especially Mg2+), glucose-1,6-diphosphate, and glutathione
  • Inhibitors include: High concentrations of chelating agents, gluconate-6-phosphate, nucleotides, and acetate
  • Can be dissolved in water (1 mg/mL) or according to specific experimental requirements
Catalog No. 50-003-98623