Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospholipase C beta 3 Antibody (YA2694) | 10 UL
SDP

Supplier:  Medchemexpress LLC HYP8294910UL

Encompass_Preferred

The Phospholipase C beta 3 Antibody (YA2694) is a non-conjugated rabbit antibody designed for detecting human phospholipase C beta 3. It is supplied as a liquid in phosphate buffered saline with sodium azide and glycerol, suitable for various laboratory applications.

  • Reactive with human samples
  • Suitable for Western Blot (WB) and Immunohistochemistry (IHC-P) applications
  • Predicted band size of 139 kDa and observed band size of 150 kDa
  • Liquid appearance with a concentration of 1.06 mg/mL
  • Formulated in phosphate buffered saline, pH 7.4, 150 mM NaCl, 0.02% sodium azide, and 50% glycerol
  • Recommended storage at -20°C for 1 year, avoiding repeated freeze/thaw cycles
  • Shipped with blue ice

Catalog No. 50-004-45086


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.