Learn More
Medchemexpress LLC Phosphonic acid, [1-(acetylamino)propylidene]bis-, calcium sodium salt (1:1:1) | 97759-16-5 | 99.0% | C5H10CaNNaO7P2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1395010MG
Encompass_Preferred
S186 is a small-molecule strontium-specific chelator provided for research use as a biochemical assay reagent (CAS 97759-16-5). It has a molecular weight of 321.15 and is supplied in milligram-scale quantities for laboratory assays; recommended storage is sealed at 4°C, with long-term storage in solvent at -80°C.
  • Strontium-specific chelation activity.
  • High purity (99.0%) suitable for research assays.
  • Available in small pack sizes for assay-scale use.
  • Stable under recommended storage conditions.
  • Molecular formula C5H10CaNNaO7P2 and molecular weight 321.15.
Catalog No. 50-002-61657