Learn More
Medchemexpress LLC Phosphonic acid, [1-(acetylamino)propylidene]bis-, calcium sodium salt (1:1:1) (9CI) | 97759-16-5 | MFCD28168001 | 99.0% | 321.15 | 25 MG

Supplier: Medchemexpress LLC 25MGHY13950
S186 is a new type of strontium-specific chelator with a molecular formula of C5H10CaNNaO7P2 and a molecular weight of 321.15. This product is intended for research use only and has not been fully validated for medical applications.
- Store at 4°C, sealed, away from moisture
- When in solvent, store at -80°C for 6 months or -20°C for 1 month
- Soluble in H2O at 25 mg/mL (77.85 mM), requiring ultrasonic
- Insoluble or slightly soluble in DMSO (< 1 mg/mL)
- Classified as a biochemical assay reagent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.