Learn More
Medchemexpress LLC Phosphonic acid, P-[[(1R)-2-(6-amino-9H-purin-9-yl)-1-methylethoxy]methyl] hydrate | 206184-49-8 | MFCD11519982 | 99.5% | C10H16N5O4P·H2O | 10MG
SDP
Supplier:  Medchemexpress LLC HY13910A10MG
Encompass_Preferred
Tenofovir hydrate is an antiviral nucleotide reverse transcriptase inhibitor provided for research use. It is used in studies of HIV and chronic hepatitis B and is supplied as a high-purity white to off-white powder suitable for biochemical and cell-based assays. This product is intended for laboratory research and not for clinical use.
  • High purity suitable for research applications.
  • White to off-white powder, easy to weigh and dissolve.
  • Available as solid, solution in DMSO, or pre-dissolved formats.
  • Suitable for biochemical and cell-based antiviral assays.
  • Batch-specific certificate of analysis available for validation.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-60772