Learn More
Medchemexpress LLC Phosphonic acid, P,P'-[1-(4-hydroxyphenoxy)ethylidene]bis- | 142523-38-4 | MFCD00673897 | 99.5% | 298.12 g·mol⁻¹ | C8H12O8P2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10107525MG
Encompass_Preferred
L-690330 is a research-grade small molecule inhibitor of inositol monophosphatase (IMPase) commonly used in biochemical and pharmacological studies. It is supplied as a hydrate in milligram quantities and is characterized by high purity and documented solubility and storage conditions.
Catalog No. 50-002-13483