Learn More
Selleck Chemical LLC Phosphonoacetic acid S3356-1g
SDP
Supplier:  Selleck Chemical LLC S33561G
Encompass_Preferred
Phosphonoacetic acid (PAA) is an active endogenous metabolite that possesses a restricted potential to block DNA biosynthesis Phosphonoacetic acid exhibits anti-viral activities
Catalog No. 50-310-9572