Learn More
Cayman Chemical PhosphoramID Mustardcycloh 1mg
SDP
Supplier:  Cayman Chemical 340781 MG
Encompass_Preferred
An alkylating agent and active metabolite of cyclophosphamide; induces DNA crosslinking, alkylates guanine in DNA, and increases the production of covalent DNA-protein conjugates in, and is cytotoxic to, HT-1080 cells; toxic to adult mice and teratogenic to embryos when administered to pregnant dams at 154 mg/kg on day 11 of gestation
Catalog No. 50-294-7715