Learn More
Carbosynth LLC. Phosphoramidon, 25mg, MW: 541.49• 45.98• 36.03, Molecular formula: C23H32N3O10P • 2 Na • 2H20, CAS# 119942-99-3
SDP
Supplier:  Carbosynth LLC. IPO408225MG
Encompass_Preferred
Synonyms: N-(α-Rhamnopyranosyloxyhydroxyphosphinyl)-L-Leucyl-L-Tryptophan • Disodium Salt Dihydrate, Application: Microbial Product Inhibitor for Thermolysin, Neutral Endopeptidase-2411 (ANP Degradation Enzyme), and Endothelin Converting Enzyme, Storage Temperature: -20°C, References: H. Suda, T. Aoyagi, T. Takeuchi, and H. Umezawa, J. Antibiotics, 26, 621 (1973). (Original), S.L. Stephenson and A.J. Kenny, J. Biochem. , 243, 183 (1987). (Pharmacol.), B.P. Roques and A. Beaumont, Trends Pharmacol. Sci., 11, 245 (1990). (Review)
Catalog No. 50-168-5895