Learn More
Enzo Life Sciences Phosphoramidon (5mg). CAS: 119942-99-3
SDP
Supplier:  Enzo Life Sciences BMLPI1110005
Encompass_Preferred
Inhibits neutral endopeptidase 24.11 (NEP, neprilysin; EC 3.4.24.11) (Ki=2nM), a cell surface zinc metalloprotease which cleaves bioactive peptides such as bradykinin, neurotensin, and enkephalin. Also inhibits neprilysin 2 (Ki~2nM). Potent inhibitor of endothelin-converting enzyme (ECE) (IC50=0.68μM). Alternative name: N-(alpha-Rhamnopyranosyloxyhydroxyphosphinyl)-L-leucyl-L-tryptophan. Purity: ≥97% (HPLC). Solubility: Soluble in water, DMSO or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-1446