Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phosphoramidon disodium | 164204-38-0 | MFCD00077870 | 99.8% | 587.47 | C23H32N3Na2O10P | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN2021A10MG

Encompass_Preferred

Phosphoramidon disodium is a microbial metabolite and metalloprotease inhibitor used as a research reagent to study metallopeptidases. It potently inhibits thermolysin (IC50 ≈ 0.4 μg/mL) and shows activity against endothelin-converting enzyme, neutral endopeptidase, and angiotensin-converting enzyme.

  • High purity: 99.8% (typical, datasheet)
  • White to off-white solid suitable for biochemical assays
  • Soluble in water (≥140 mg/mL) and DMSO (100 mg/mL; sonication recommended)
  • Storage: store sealed at 4°C; in solution store -80°C (up to 6 months) or -20°C (up to 1 month)
  • Supplied in small research quantities, typically 10 mg

Catalog No. 50-002-95580


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.