Learn More
Crescent Chemical Co Inc PHOSPHORAMIDON RESEARCH GRADE
SDP
Supplier:  Crescent Chemical Co Inc SE32753.01
Encompass_Preferred
Phosphoramidon research grade Inhibitor for thermolysin and neutral endopeptidase-24.11 (ANP Degradation Enzyme). Inhibits the activity of 'Endothelin Converting Enzyme' (3 - 9). Microbial product.Store dry and protect from light! Size - 5MG Storage Con
Catalog No. 50-248-016