Learn More
Medchemexpress LLC Phosphoric acid, 2,2-bis(chloromethyl)-1,3-propanediyl tetrakis(2-chloroethyl) ester | 38051-10-4 | 583.0 g/mol | C13H24Cl6O8P2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW58582610MG
Encompass_Preferred
Antiblaze 100 (Antiblaze V6) is a chlorinated organophosphate additive flame retardant (CAS 38051-10-4) used historically in flexible polyurethane foam and other polymer formulations. Its molecular formula is C13H24Cl6O8P2 and molecular weight is 583.0 g/mol. The substance is documented in regulatory and safety literature and may contain chlorinated impurities; handle and dispose of it according to the SDS and applicable regulations.
  • Additive flame retardant for flexible polyurethane foam
  • Chlorinated organophosphate chemical class
  • Low-volatility, oil-like liquid suitable for polymer incorporation
  • Documented in regulatory and safety assessments
  • Requires SDS-guided handling and disposal
Catalog No. 50-002-84541