Learn More
Medchemexpress LLC Phosphoric acid tributyl ester-d27 | 61196-26-7 | 100.0% | 293.48 | C12D27O4P | 50 MG
SDP
Supplier:  Medchemexpress LLC HY117600S50MG
Encompass_Preferred
Deuterium-labeled tributyl phosphate (Tributyl phosphate-d27) supplied as a 50 mg solid for research use. It is intended as an internal standard and tracer for analytical techniques including NMR, GC-MS, and LC-MS, and is suitable for quantitative and structural studies. Store and handle under recommended conditions to preserve isotopic and chemical integrity.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS
  • High isotopic purity for accurate quantitation
  • Soluble in DMSO at ≥100 mg/mL for convenient stock preparation
  • Suitable for use as a tracer or internal standard in analytical workflows
  • Stable as a solid when stored at -20°C under recommended conditions
Catalog No. 50-002-26676