Learn More
Strem, An Ascensus Company CAS 167709-31-1. 10g. (S)-(-)-22-Bis[di(35-di-t-butylphenyl)phosphino]-66-dimethoxy-11-biphenyl min. 97%. MFCD09753001
SDP
Supplier:  Strem, An Ascensus Company 15004510G
Encompass
CAS 167709-31-1. 10g. (S)-(-)-22-Bis[di(35-di-t-butylphenyl)phosphino]-66-dimethoxy-11-biphenyl min. 97%. MFCD09753001. Molecular Weight 1031.49. Molecular Formula C70H96O2P2. Color/form white pwdr. Strem 15-0045. http//www.strem.com/catalog/v/15-0045/
Catalog No. 50-211-0721