Learn More
CPC Scientific H-Lys-Arg-Asn-Pro-Gly-Ser-Gln-Lys-Arg-Phe-Pro-Ser-Asn-Cys-Gly-Arg-Asp-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP027B
Encompass
ONE-LETTER SEQUENCE: KRNPGSQKRFPSNCGRDMOLECULAR FORMULA: C79H131N31O25S1MOLECULAR WEIGHT:1947.14STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [150829-21-3], RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: V.E. Sanchez et al., J. Bio. Chem., 268, 17889 (1993)
Catalog No. 50-211-0178