Learn More
Medchemexpress LLC HY-P7407 10ug Medchemexpress, Sonic Hedgehog, C24II, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP740710UG
Encompass_Preferred
Medchemexpress, HY-B2233 500mg Phosphorylcholine (chloride) CAS:107-73-3 Phosphorylcholine chloride (Phosphocholine chloride) is an antigenic cell-surface component found on many commensal and pathogenic bacteria that reside in the upper airway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0611