Learn More
Medchemexpress LLC HY-B2233B 500mg Medchemexpress, Phosphorylcholine CAS:3616-04-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2233B500MG
Encompass_Preferred
Medchemexpress, HY-B2233B 500mg Phosphorylcholine CAS:3616-04-4 Phosphatidylcholine is the main phospholipid component in eukaryotic biofilms. Phosphatidylcholine exists in commensal or pathogenic bacteria associated with eukaryotes in prokaryotes. Phosphorylcholine exhibits a surprising range of immunomodulatory properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0179