Learn More
Medchemexpress LLC Phosphorylcholine chloride | 107-73-3 | 99.95% | 219.60 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW011249100G
Encompass_Preferred
Phosphorylcholine chloride is identified as the primary phospholipid component within eukaryotic biofilms, also found in prokaryotes associated with eukaryotes, including commensal or pathogenic bacteria. This compound demonstrates a broad spectrum of immunomodulatory characteristics.
  • Acts as a component of drug delivery micelles.
  • Enhances the efficiency of cellular internalization.
  • Boosts the cytotoxicity of cytotoxins delivered by micelles in HeLa cells.
  • Activates nicotinic and muscarinic cholinergic neurotransmission.
  • Stimulates the activation of α2-adrenergic receptors.
  • Elevates plasma glucagon and choline concentrations.
  • Promotes peripheral autonomic nervous system activity.
  • Induces the creation of anti-PC and anti-oxLDL antibodies.
  • Increases mature b cells in the spleen.
  • Decreases atherosclerotic lesions in the aortic root in mouse models.
Catalog No. 50-004-51884