Learn More
Meso Scale Discovery UPLEX HUM GRO ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151UXK4
Encompass
The Meso Scale Discovery U-PLEX Human GRO-alpha Assay Kit is a multiplex immunoassay that measures GRO-alpha levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It allows simultaneous measurement of GRO-alpha and other biomarkers The assay takes 3 5 hours to complete after a 2 hour incubation
Catalog No. 50-000-80530
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.