Learn More
Meso Scale Discovery PhosphoSTAT5ab WCL
SDP
Supplier:  Meso Scale Discovery K150IGD1
Encompass
The Meso Scale Discovery Phospho-STAT5a/b Whole Cell Lysate Kit is designed to measure endogenous levels of STAT5a/b protein phosphorylated at Tyr694/Tyr699 in cell lysates The kit includes a 96-well plate coated with capture antibody a detection antibody conjugated to MSD SULFO-TAG and cell lysis buffer The electrochemiluminescence assay allows for high sensitivity and a broad dynamic range This kit can be used to monitor STAT5 phosphorylation in response to cytokines and growth factors in a variety of cell lines The kit provides all necessary reagents for cell lysis and detection in a convenient format
Catalog No. 50-000-81702