Learn More
Hach Company PhosVer 3 HHC Reagent Set
Supplier:  Hach Company 2135100
Encompass
This reagent set was developed specifically for customers who use Phosver 3 in high humidity regions For determination of reactive (ortho) phosphate (phosphorus) by the PhosVer 3 ascorbic acid method using Hach Method 10318 Sample Size 10 mL 100 samples
Catalog No. 50-002-56963