Learn More
Perkin Elmer US LLC Holmium Oxide Glass Filter
Supplier:  Perkin Elmer US LLC B0080832
Encompass_Preferred
Photometric and Wavelength Accuracy Reference standards are a set of standards used to verify the ordinate (photometric) and abscissa (wavelength) accuracy of UV/Vis and UV/Vis/NIR spectrometers. The neutral density glass filters are A2LA ISO 17025 accredited.
Catalog No. 50-201-6385