Learn More
ABclonal Technology PHPT1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1359020UL
Encompass_Preferred
This gene encodes an enzyme that catalyzes the reversible dephosphorylation of histidine residues in proteins. It may be involved in the dephosphorylation of G-beta and ATP citrate lyase and in negatively regulating CD4 T lymphocytes by dephosphorylation and inhibition of KCa3.1 channels. Alternative splicing results in multiple transcript variants.
Catalog No. 50-000-97426