Learn More
STA PHARMACEUTICAL US LLC PHT PEG3 amidite | 1 g | CAS 2170381-91-4 | InChIKey UDTLXDXACDJZQQ-UHFFFAOYSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10686MM301GM
PHT PEG3 amidite is a Amidite reagent (Subcategory: Amino Modifiers) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: 2170381-91-4
- MDL: No data
- InChIKey: UDTLXDXACDJZQQ-UHFFFAOYSA-N
- Molecular Weight: 495.512761998
- Molecular Formula: C23H34N3O7P
- Purity: ≥95%
- Container Type: 30 mL Glass (28-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 50.1 g
- Commodity Code: 29280090
- Country Of Origin: China
- IUPAC: 2-cyanoethyl (2-(2-(2-((1,3-dioxoisoindolin-2-yl)oxy)ethoxy)ethoxy)ethyl) diisopropylphosphoramidite
- SMILES: CC(C)N(C(C)C)P(OCCOCCOCCON1C(C2=C(C=CC=C2)C1=O)=O)OCCC#N
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.